Please login to your online account to display your discounted pricing

2-Cyano-6-methoxybenzothiazole, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  225360010

 View more versions of this product

Catalog No. AC225360010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Formula Weight 190.23
Assay ≥98.5%
Quantity 1g
Color Off-white to light yellow
Melting Point 128° to 131°C
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 2-Cyano-6-methoxybenzothiazole
Molecular Formula C9H6N2OS
CAS 943-03-3
Assay Percent Range 99%
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 98.5
MDL Number MFCD00010537
Infrared Spectrum Authentic
Synonym 6-Methoxy-2-benzothiazolecarbonitrile
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

Specifications

Chemical Name or Material 2-Cyano-6-methoxybenzothiazole
CAS 943-03-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H6N2OS
Formula Weight 190.23
MDL Number MFCD00010537
Synonym 6-Methoxy-2-benzothiazolecarbonitrile
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.

SDS & Safety Documents