Learn More
2-Cyano-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6448603
Description
It is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a useful reactant in the synthesis of s-tetrazines. Also act as a intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
2-Cyano-5-(trifluoromethyl)pyridine | |
C7H3F3N2 | |
1 g | |
13339826 | |
Slightly soluble in water. | |
FC(F)(F)C1=CN=C(C=C1)C#N | |
172.11 | |
172.11 | |
Solid |
95727-86-9 | |
MFCD01001119 | |
UN3439 | |
5-trifluoromethyl picolinonitrile, 5-trifluoromethyl pyridine-2-carbonitrile, 2-cyano-5-trifluoromethyl pyridine, 2-cyano-5-trifluoromethylpyridine, 5-trifluoromethyl-2-pyridinecarbonitrile, 5-trifluoromethyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 5-trifluoromethyl, pubchem17518, 5-trifluoromethyl ;picolinonitrile, 2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile | |
WDSCJULUXJSJOX-UHFFFAOYSA-N | |
5-(trifluoromethyl)pyridine-2-carbonitrile | |
1535372 | |
98% |
Chemical Identifiers
95727-86-9 | |
172.11 | |
WDSCJULUXJSJOX-UHFFFAOYSA-N | |
1535372 | |
FC(F)(F)C1=CN=C(C=C1)C#N |
C7H3F3N2 | |
MFCD01001119 | |
5-trifluoromethyl picolinonitrile, 5-trifluoromethyl pyridine-2-carbonitrile, 2-cyano-5-trifluoromethyl pyridine, 2-cyano-5-trifluoromethylpyridine, 5-trifluoromethyl-2-pyridinecarbonitrile, 5-trifluoromethyl-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 5-trifluoromethyl, pubchem17518, 5-trifluoromethyl ;picolinonitrile, 2-cyano-5-trifluoromethyl pyridine, 5-trifluoromethyl picolinonitrile | |
5-(trifluoromethyl)pyridine-2-carbonitrile |
Safety and Handling
GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only