Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyano-5-fluoropyridine, 98+%

$83.93 - $789.54

Chemical Identifiers

Chemical Name or Material 2-Cyano-5-fluoropyridine
CAS 327056-62-2
Molecular Formula C6H3FN2
Formula Weight 122.1
MDL Number MFCD08741371
Synonym 5-Fluoropicolinonitrile; 5-Fluoropyridine-2-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2595603

  SDS
alfa aesar™
H2595603
1g Each for $83.93

AAH2595606

  SDS
alfa aesar™
H2595606
5g Each for $250.12

AAH2595614

  SDS
alfa aesar™
H2595614
25g Each for $789.54
Description & Specifications

Specifications

Formula Weight 122.1
Assay ≥98%
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 327056-62-2
MDL Number MFCD08741371
Synonym 5-Fluoropicolinonitrile; 5-Fluoropyridine-2-carbonitrile
Chemical Name or Material 2-Cyano-5-fluoropyridine
Molecular Formula C6H3FN2
Melting Point 41° to 44°C
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.