Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyano-4-(trifluoromethyl)benzeneboronic acid, 95%

Chemical Identifiers

Chemical Name or Material 2-Cyano-4-(trifluoromethyl)benzeneboronic acid
CAS 1218790-84-1
Formula Weight 214.94
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6145103

 
alfa aesar™
H6145103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61451MD

 
alfa aesar™
H61451MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.94
Chemical Name or Material 2-Cyano-4-(trifluoromethyl)benzeneboronic acid
Physical Form Solid
Assay 95%
CAS 1218790-84-1
Quantity 1g