missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Cyano-3-methylpyridine, 98%, ACROS Organics™

Chemical Identifiers

CAS 20970-75-6
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.139
MDL Number MFCD00190585
InChI Key WBXZCDIZXWDPBL-UHFFFAOYSA-N
Synonym 2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
PubChem CID 819928
IUPAC Name 3-methylpyridine-2-carbonitrile
SMILES CC1=C(N=CC=C1)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC345520050 SDS View Documents Acros Organics
345520050
5g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC345521000 SDS View Documents Acros Organics
AC345521000
100g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

20970-75-6
118.139
WBXZCDIZXWDPBL-UHFFFAOYSA-N
819928
CC1=C(N=CC=C1)C#N
C7H6N2
MFCD00190585
2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
3-methylpyridine-2-carbonitrile

Specifications

Authentic
Beige
5g
97.5
98%
MFCD00190585
WBXZCDIZXWDPBL-UHFFFAOYSA-N
3-methylpyridine-2-carbonitrile
819928
Crystalline Powder
2-Cyano-3-methylpyridine, 98%
Glass bottle
83°C to 87°C
20970-75-6
100.0
C7H6N2
2-cyano-3-methylpyridine, 3-methylpicolinonitrile, 2-cyano-3-methyl pyridine, 3-methyl-2-pyridinecarbonitrile, pyridinecarbonitrile, methyl, 3-methyl-pyridine-2-carbonitrile, 2-cyano-3-methyl-pyridine, 2-pyridinecarbonitrile, 3-methyl, 2-cyano-3-methylpyridine 2-cyano-3-picoline, 2-cyano-3-picoline
CC1=C(N=CC=C1)C#N
118.139
118.14
≥97.5%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 244-131-3

TSCA : TSCA