Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyano-3-methoxybenzeneboronic acid, 95%

Chemical Identifiers

Chemical Name or Material 2-Cyano-3-methoxybenzeneboronic acid
CAS 1164100-84-8
Formula Weight 176.97
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH61856MC

 
alfa aesar™
H61856MC
100mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 176.97
Chemical Name or Material 2-Cyano-3-methoxybenzeneboronic acid
Quantity 100mg
CAS 1164100-84-8
Physical Form Solid
Assay 95%