Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyano-3,5-difluoropyridine, 98%

$68.70 - $207.93

Chemical Identifiers

Chemical Name or Material 2-Cyano-3,5-difluoropyridine
CAS 298709-29-2
Molecular Formula C6H2F2N2
Formula Weight 140.09
MDL Number MFCD03788759
Synonym 3,5-Difluoropicolinitrile; 3,5-Difluoropyridine-2-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1972403

  SDS
alfa aesar™
L1972403
1g Each for $68.70

AAL1972406

  SDS
alfa aesar™
L1972406
5g Each for $207.93
Description & Specifications

Specifications

Formula Weight 140.09
Chemical Name or Material 2-Cyano-3,5-difluoropyridine
Molecular Formula C6H2F2N2
CAS 298709-29-2
Health Hazard 2 GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Assay 98%
MDL Number MFCD03788759
Synonym 3,5-Difluoropicolinitrile; 3,5-Difluoropyridine-2-carbonitrile
Health Hazard 1 Warning
Melting Point 30° to 34°C
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.