Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Cyano-2,2-dimethylacetic acid, 95%

Chemical Identifiers

Chemical Name or Material 2-Cyano-2,2-dimethylacetic acid
CAS 22426-30-8
Formula Weight 113.11
MDL Number MFCD11100210
UN Number UN2923
Synonym 2-Cyano-2-methylpropionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6673403

  SDS
alfa aesar™
H6673403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66734MD

  SDS
alfa aesar™
H66734MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 56° to 61°C
Formula Weight 113.11
Boiling Point 132° to 135°C (12mmHg)
CAS 22426-30-8
Physical Form Crystalline Solid
Chemical Name or Material 2-Cyano-2,2-dimethylacetic acid
Assay 95%
MDL Number MFCD11100210
Synonym 2-Cyano-2-methylpropionic acid
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Health Hazard 1 Danger
Quantity 1g
Health Hazard 2 GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
UN Number UN2923
Color White
Sensitivity Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.