missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroveratraldehyde, 98%

$96.24 - $371.65

Chemical Identifiers

CAS 5417-17-4
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00084942
InChI Key SAWHDJTZESXNMM-UHFFFAOYSA-N
Synonym 2-chloroveratraldehyde, 2-chloro-3,4-dimethoxy-benzaldehyde, benzaldehyde, 2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxy benzaldehyde, chloroveratraldehyde, pubchem7759, acmc-1aumn, 2-chloro-3,4 dimethoxybenzaldehyde, benzaldehyde,2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxybenzaldehyde
PubChem CID 79445
IUPAC Name 2-chloro-3,4-dimethoxybenzaldehyde
SMILES COC1=C(C(=C(C=C1)C=O)Cl)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AAA1062306 missing translation for 'viewDocuments' Alfa Aesar™
A1062306
5g Each for $96.24
 
AAA1062314 missing translation for 'viewDocuments' Alfa Aesar™
A1062314
25g Each for $371.65
 
Specifications

Chemical Identifiers

5417-17-4
200.618
SAWHDJTZESXNMM-UHFFFAOYSA-N
79445
COC1=C(C(=C(C=C1)C=O)Cl)OC
C9H9ClO3
MFCD00084942
2-chloroveratraldehyde, 2-chloro-3,4-dimethoxy-benzaldehyde, benzaldehyde, 2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxy benzaldehyde, chloroveratraldehyde, pubchem7759, acmc-1aumn, 2-chloro-3,4 dimethoxybenzaldehyde, benzaldehyde,2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxybenzaldehyde
2-chloro-3,4-dimethoxybenzaldehyde

Specifications

70°C to 72°C
5417-17-4
MFCD00084942
2-chloroveratraldehyde, 2-chloro-3,4-dimethoxy-benzaldehyde, benzaldehyde, 2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxy benzaldehyde, chloroveratraldehyde, pubchem7759, acmc-1aumn, 2-chloro-3,4 dimethoxybenzaldehyde, benzaldehyde,2-chloro-3,4-dimethoxy, 2-chloro-3,4-dimethoxybenzaldehyde
COC1=C(C(=C(C=C1)C=O)Cl)OC
200.618
200.62
2-Chloroveratraldehyde
5g
C9H9ClO3
2109344
SAWHDJTZESXNMM-UHFFFAOYSA-N
2-chloro-3,4-dimethoxybenzaldehyde
79445
98%