Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorotrityl alcohol on polystyrene, 1% cross-linked, 100-200 mesh, 0.6-1.5 mmol/g

$461.78 - $461.78

Chemical Identifiers

Chemical Name or Material 2-Chlorotrityl alcohol on polystyrene
Name Note 1% cross-linked
MDL Number MFCD02683514
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1960206

 
alfa aesar™
L1960206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1960214

 
alfa aesar™
L1960214
25g Each for $461.78
Description & Specifications

Specifications

Chemical Name or Material 2-Chlorotrityl alcohol on polystyrene
Mesh Size 100 to 200 mesh
MDL Number MFCD02683514
Concentration 0.6 to 1.5mmol/g
Name Note 1% cross-linked
Quantity 5g