2-Chlorotrityl alcohol on polystyrene, 1% cross-linked, 100-200 mesh, 0.6-1.5 mmol/g, Thermo Scientific™
Supplier: Thermo Scientific Chemicals L1960206
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Chlorotrityl alcohol on polystyrene | |
MFCD02683514 | |
100-200 Mesh |
1% cross-linked | |
5g |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO â Research Use Only