Please login to your online account to display your discounted pricing

2-Chlorotoluene, 98%, ACROS Organics™

$15.30 - $78.75

Chemical Identifiers

Chemical Name or Material 2-Chlorotoluene
CAS 95-49-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7Cl
Linear Formula CH3C6H4Cl
Formula Weight 126.59
MDL Number MFCD00000562
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC150200025

  SDS
thermo fisher scientific
AC150200025
Glass bottle 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC150200010

  SDS
thermo fisher scientific
150200010
Glass bottle 1L Each for $62.63

AC150200050

  SDS
thermo fisher scientific
150200050
Glass bottle 5mL Each for $15.30

AC150205000

  SDS
thermo fisher scientific
150205000
Glass bottle 500mL Each for $78.75
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 126.59
Quantity 2.5L
Color Clear
Density 1.0800g/mL
Beilstein 05, 290
Merck Index 15, 2175
CAS Max % 100.0
Chemical Name or Material 2-Chlorotoluene
Water 0.015% max.
Assay Percent Range 98%
Molecular Formula C7H7Cl
CAS 95-49-8
Packaging Glass bottle
Refractive Index 1.5240 to 1.5260 (20°C, 589nm)
Linear Formula CH3C6H4Cl
MDL Number MFCD00000562
Synonym 1-Chloro-2-methylbenzene
CAS Min % 97.5
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Suspected of damaging the unborn child.
Health Hazard 3 GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Obtain special instructions before use. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Warning

GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Obtain special instructions before use. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Harmful if inhaled.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
Suspected of damaging the unborn child.