missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chlorothioxanthone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1813106
Description
2-Chlorothioxanthone is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorothioxanthone is used as pharmaceutical intermediate.
Solubility
Partly miscible in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
2-Chlorothioxanthone | |
151°C to 154°C | |
C13H7ClOS | |
5 g | |
Partly miscible in water. | |
ClC1=CC=C2SC3=CC=CC=C3C(=O)C2=C1 | |
246.71 | |
246.72 |
86-39-5 | |
91°C (196°F) | |
MFCD00005067 | |
2-chlorothioxanthone, 2-chloro-9h-thioxanthen-9-one, 9h-thioxanthen-9-one, 2-chloro, unii-rgs87t004b, 2-chlorothiaxanthone, 2-chlorothioxanthene-9-one, sandoray 1050, 2-chloro-thioxanthone, pubchem10692, 2-chloro-9-thioxanthone | |
ZCDADJXRUCOCJE-UHFFFAOYSA-N | |
2-chloro-9H-thioxanthen-9-one | |
618848 | |
99% |
Chemical Identifiers
86-39-5 | |
246.71 | |
ZCDADJXRUCOCJE-UHFFFAOYSA-N | |
618848 | |
ClC1=CC=C2SC3=CC=CC=C3C(=O)C2=C1 |
C13H7ClOS | |
MFCD00005067 | |
2-chlorothioxanthone, 2-chloro-9h-thioxanthen-9-one, 9h-thioxanthen-9-one, 2-chloro, unii-rgs87t004b, 2-chlorothiaxanthone, 2-chlorothioxanthene-9-one, sandoray 1050, 2-chloro-thioxanthone, pubchem10692, 2-chloro-9-thioxanthone | |
2-chloro-9H-thioxanthen-9-one |
Safety and Handling
EINECSNumber : 201-667-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only