missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chlorothiophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC224350250

 View more versions of this product

Catalog No. AC224350250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-chlorothiophenol
Clear slightly yellow
1614434
100.0
Glass bottle
Liquid
98%
ClC6H4SH
MFCD00004830
1.275g/mL
25g
97.5
Authentic
1.6020 to 1.6040 (20°C, 589nm)
≥97.5%
C6H5ClS
144.62
2-chlorobenzenethiol

Chemical Identifiers

1614434
100.0
C6H5ClS
144.62
2-chlorobenzenethiol
≥97.5%
97.5
98%
ClC6H4SH
MFCD00004830
2-chlorothiophenol

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger