missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorothiophene, 98+%

$60.77 - $738.68

Chemical Identifiers

CAS 96-43-5
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.578
MDL Number MFCD00005421
InChI Key GSFNQBFZFXUTBN-UHFFFAOYSA-N
Synonym 2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
PubChem CID 7299
IUPAC Name 2-chlorothiophene
SMILES C1=CSC(=C1)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA103900B View Documents Alfa Aesar™
A103900B
1000g Each for $738.68
 
AAA1039030 View Documents Alfa Aesar™
A1039030
250g Each for $234.41
 
AAA1039018 View Documents Alfa Aesar™
A1039018
50g Each for $60.77
 
Description

Description

Applications
It is used in the preparation of 5-phenylthiophene derivative and N-arylhydroxamic acids.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
Specifications

Chemical Identifiers

96-43-5
118.578
GSFNQBFZFXUTBN-UHFFFAOYSA-N
7299
C1=CSC(=C1)Cl
C4H3ClS
MFCD00005421
2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
2-chlorothiophene

Specifications

22°C (71°F)
127°C to 129°C
1000g
C4H3ClS
UN1992
2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
C1=CSC(=C1)Cl
118.578
118.59
2-Chlorothiophene
Unpleasant
-72°C
96-43-5
MFCD00005421
104652
GSFNQBFZFXUTBN-UHFFFAOYSA-N
2-chlorothiophene
7299
≥98%
Safety and Handling

Safety and Handling

GHS H Statement
H225-H301-H311-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.

TSCA : TSCA