Please login to your online account to display your discounted pricing

2-Chlorothiophene, 97%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  168261000

 View more versions of this product

Catalog No. AC168261000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥96.0%
Formula Weight 118.58
Quantity 100mL
Color Clear colorless to light brown
Beilstein 17, 32
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Chlorothiophene
Packaging Glass bottle
Storage Information Product darkens on storage or exposure to light
Assay Percent Range 97%
Molecular Formula C4H3ClS
MDL Number MFCD00005421
CAS 96-43-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.5470 to 1.5490 (20°C, 589nm)
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Chlorothiophene
CAS 96-43-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H3ClS
Formula Weight 118.58
MDL Number MFCD00005421
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.

SDS & Safety Documents