Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorothioisonicotinamide, 97%

Chemical Identifiers

Chemical Name or Material 2-Chlorothioisonicotinamide
Molecular Formula C6H5ClN2S
Formula Weight 172.63
MDL Number MFCD10686940
Synonym 2-Chloropyridine-4-thiocarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5226306

  SDS
alfa aesar™
H5226306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.63
Chemical Name or Material 2-Chlorothioisonicotinamide
Assay 97%
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Molecular Formula C6H5ClN2S
Synonym 2-Chloropyridine-4-thiocarboxamide
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
MDL Number MFCD10686940
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.