missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloroquinoline, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 110040050

 View more versions of this product

Catalog No. AC110040050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Chloroquinoline
>110°C
Glass bottle
265°C to 267°C
34°C to 37°C
612-62-4
100.0
C9H6ClN
20, 359
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,benzene,petrol
C1=CC=C2C(=C1)C=CC(=N2)Cl
163.61
163.61
99%
1.2300g/mL
Authentic
1.23
White to Yellow
5g
98.5
98.5% min. (GC)
MFCD00006741
2-chloro quinoline, 2-chloro-quinoline, ccris 3977, chioroquinoline, chloroquinoline, pubchem7580, quinoline, 2-chloro, quinoline, chloro, unii-ux7rin8gew, ux7rin8gew
OFUFXTHGZWIDDB-UHFFFAOYSA-N
2-chloroquinoline
11928
Crystalline Powder, Crystals and/or Chunks or Fused Mass

Chemical Identifiers

612-62-4
163.61
OFUFXTHGZWIDDB-UHFFFAOYSA-N
11928
C1=CC=C2C(=C1)C=CC(=N2)Cl
C9H6ClN
MFCD00006741
2-chloro quinoline, 2-chloro-quinoline, ccris 3977, chioroquinoline, chloroquinoline, pubchem7580, quinoline, 2-chloro, quinoline, chloro, unii-ux7rin8gew, ux7rin8gew
2-chloroquinoline
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 210-317-8

SDS Documents
Product Certifications