Please login to your online account to display your discounted pricing

2-chloroquinoline, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  110040010

 View more versions of this product

Catalog No. AC110040010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 163.61
Assay ≥98.5%
Physical Form Crystalline Powder, Crystals and/or Chunks or Fused Mass
Quantity 1g
Color White to yellow
Beilstein 20, 359
Melting Point 34° to 38°C
Density 1.2300g/mL
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 Warning
MDL Number MFCD00006741
Assay Percent Range 99%
CAS Min % 98.5
Infrared Spectrum Authentic
Molecular Formula C9H6ClN
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 612-62-4
Chemical Name or Material 2-Chloroquinoline

Specifications

Chemical Name or Material 2-Chloroquinoline
CAS 612-62-4
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H6ClN
Formula Weight 163.61
MDL Number MFCD00006741
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents