Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroquinoline, 99%

$134.69 - $387.89

Chemical Identifiers

Chemical Name or Material 2-Chloroquinoline
CAS 612-62-4
Molecular Formula C9H6ClN
Formula Weight 163.61
MDL Number MFCD00006741
Beilstein 112561
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2344306

  SDS
alfa aesar™
B2344306
5g Each for $134.69

AAB2344314

  SDS
alfa aesar™
B2344314
25g Each for $387.89
Description & Specifications

Specifications

Formula Weight 163.61
Chemical Name or Material 2-Chloroquinoline
TSCA TSCA
Assay 99%
MDL Number MFCD00006741
CAS 612-62-4
Beilstein 112561
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
RTECS Number VB2320000
Flash Point >110°C (230°F)
Odor Characteristic
Boiling Point 265° to 267°C
Melting Point 34° to 37°C
Quantity 5g
Molecular Formula C9H6ClN

Applications
2-Chloroquinoline is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. It possess antimicrobial activity against a panel of bacterial and fungal strains. It reacts with benzotriazole, followed by cyclization with loss of nitrogen, gives the indolo[2,3-b]quinoline ring system for use in novel cytotoxic DNA topoisomerase II inhibitors.

Solubility
Soluble in ethanol, ether, methanol, and chloroform. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.