Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroquinoline-3-carboxaldehyde, 98%

$46.09 - $206.43

Chemical Identifiers

Chemical Name or Material 2-Chloroquinoline-3-carboxaldehyde
CAS 73568-25-9
Molecular Formula C10H6ClNO
Formula Weight 191.62
MDL Number MFCD00130079
Synonym 2-Chloro-3-formylquinoline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2713303

  SDS
alfa aesar™
H2713303
1g Each for $46.09

AAH2713309

  SDS
alfa aesar™
H2713309
10g Each for $206.43
Description & Specifications

Specifications

Formula Weight 191.62
Synonym 2-Chloro-3-formylquinoline
Chemical Name or Material 2-Chloroquinoline-3-carboxaldehyde
Melting Point 148° to 150°C
Quantity 1g
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.
MDL Number MFCD00130079
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Molecular Formula C10H6ClNO
CAS 73568-25-9
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338-P337+P313
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.