Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroquinoline-3-boronic acid, 97%

$222.07 - $222.07

Chemical Identifiers

Chemical Name or Material 2-Chloroquinoline-3-boronic acid
CAS 128676-84-6
Molecular Formula C9H7BClNO2
Formula Weight 207.43g/mol
MDL Number MFCD01114675
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2032906

 
alfa aesar™
L20329-06
5g Each for $222.07
Description & Specifications

Specifications

Quantity 5g
Melting Point 144° to 147°C
Quantity 5g
Chemical Name or Material 2-Chloroquinoline-3-boronic acid
CAS 128676-84-6
Assay 97%
Molecular Formula C9H7BClNO2
Formula Weight 207.43g/mol
MDL Number MFCD01114675
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.