missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloropyrimidine, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 185840100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloropyrimidine | |
62.0°C to 66.0°C | |
75.0°C to 76.0°C (10.0 mmHg) | |
Authentic | |
Glass bottle | |
MFCD00006060 | |
pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin | |
ClC1=NC=CC=N1 | |
114.53 | |
114.53 | |
Crystalline Powder or Crystals |
1722-12-9 | |
Orange to Yellow | |
98°C | |
98% min. (GC) | |
C4H3ClN2 | |
10 g | |
UNCQVRBWJWWJBF-UHFFFAOYSA-N | |
2-chloropyrimidine | |
74404 | |
98+% |
Chemical Identifiers
1722-12-9 | |
114.53 | |
UNCQVRBWJWWJBF-UHFFFAOYSA-N | |
74404 | |
ClC1=NC=CC=N1 |
C4H3ClN2 | |
MFCD00006060 | |
pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin | |
2-chloropyrimidine |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 217-020-2
RUO – Research Use Only