Learn More
2-chloropyrimidine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 438450010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloropyrimidine-5-carboxaldehyde | |
96.0 | |
113°C to 118°C | |
Authentic | |
Glass bottle | |
MFCD10696891 | |
2-chloropyrimidine-5-carboxyaldehyde, 2-chloropyrimidine-5-carboxaldehyde, 2-chloro-5-pyrimidinecarboxaldehyde, pubchem23896, 2-chloro-5-formyl pyrimidine, chloropyrimidine-5-carbaldehyde, 2-chloro-pyrimidine-5-carbaldehyde, 2-chloranylpyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 2-chloro | |
ClC1=NC=C(C=O)C=N1 | |
142.54 | |
142.55 | |
Crystalline Powder or Powder |
933702-55-7 | |
100.0 | |
White to Yellow | |
96% min. (GC) | |
C5H3ClN2O | |
1 g | |
LJYQVOPFBNMTKJ-UHFFFAOYSA-N | |
2-chloropyrimidine-5-carbaldehyde | |
21698350 | |
97% |
Chemical Identifiers
933702-55-7 | |
142.54 | |
LJYQVOPFBNMTKJ-UHFFFAOYSA-N | |
21698350 | |
ClC1=NC=C(C=O)C=N1 |
C5H3ClN2O | |
MFCD10696891 | |
2-chloropyrimidine-5-carboxyaldehyde, 2-chloropyrimidine-5-carboxaldehyde, 2-chloro-5-pyrimidinecarboxaldehyde, pubchem23896, 2-chloro-5-formyl pyrimidine, chloropyrimidine-5-carbaldehyde, 2-chloro-pyrimidine-5-carbaldehyde, 2-chloranylpyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 2-chloro | |
2-chloropyrimidine-5-carbaldehyde |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw
GHS Signal Word: Warning
RUO – Research Use Only