missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyrimidine-5-boronic acid pinacol ester, 98%

Manufacturer:  Alfa Aesar™ H64052MD

 View more versions of this product

Catalog No. AAH64052MD



Specifications

Specifications

2-Chloropyrimidine-5-boronic acid pinacol ester
1003845-08-6
MFCD11856048
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidin-5-ylboronic acid pinacol ester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid, pinacol ester, 2-chloropyrimidin-5-yl boronic acid pinacol ester, 2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)Cl
240.494
240.49
98%
250mg
C10H14BClN2O2
15622160
VLAPDEKXZLRRKV-UHFFFAOYSA-N
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
51341943
Solid

Chemical Identifiers

1003845-08-6
240.494
VLAPDEKXZLRRKV-UHFFFAOYSA-N
51341943
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)Cl
C10H14BClN2O2
MFCD11856048
2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid pinacol ester, 2-chloro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2-chloropyrimidin-5-ylboronic acid pinacol ester, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidine, 2-chloropyrimidine-5-boronic acid, pinacol ester, 2-chloropyrimidin-5-yl boronic acid pinacol ester, 2-5-chloro-2-pyrimidyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-chloranyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, pyrimidine, 2-chloro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents