Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyridine N-oxide, 97%

Chemical Identifiers

Chemical Name or Material 2-Chloropyridine N-oxide
CAS 2402-95-1
Molecular Formula C5H4ClNO
Formula Weight 129.55g/mol
MDL Number MFCD00129036
UN Number UN2811
Beilstein 108585
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2003303

 
alfa aesar™
L20033-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2003306

 
alfa aesar™
L20033-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 67° to 72°C
Quantity 1g
Chemical Name or Material 2-Chloropyridine N-oxide
CAS 2402-95-1
Assay 97%
Molecular Formula C5H4ClNO
Formula Weight 129.55g/mol
MDL Number MFCD00129036
UN Number UN2811
Beilstein 108585
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Applications
2-Chloropyridine N-oxide is important intermediate products for example in the preparation of pyrithiene, especially zinc pyrithione

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers

Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C