missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyridine-5-carboxaldehyde, 96%

Manufacturer:  Alfa Aesar™ H55550MD

 View more versions of this product

Catalog No. AAH55550MD



Description

Description

Applications
2-Chloropyridine-5-carboxaldehyde is used as pharmaceutical intermediates

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

2-Chloropyridine-5-carboxaldehyde
250mg
C6H4ClNO
2-chloro-5-formylpyridine
C1=CC(=NC=C1C=O)Cl
141.554
141.56
77°C to 81°C
23100-12-1
MFCD03095223
AFWWKZCPPRPDQK-UHFFFAOYSA-N
6-chloropyridine-3-carbaldehyde
2764053
96%

Chemical Identifiers

23100-12-1
141.554
AFWWKZCPPRPDQK-UHFFFAOYSA-N
2764053
C1=CC(=NC=C1C=O)Cl
C6H4ClNO
MFCD03095223
2-chloro-5-formylpyridine
6-chloropyridine-3-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents