Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyridine-5-boronic acid pinacol ester, 98%

$123.86 - $516.82

Chemical Identifiers

Chemical Name or Material 2-Chloropyridine-5-boronic acid pinacol ester
CAS 444120-94-9
Molecular Formula C11H15BClNO2
Formula Weight 239.51
MDL Number MFCD03412793
Synonym 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5007103

  SDS
alfa aesar™
H5007103
1g Each for $123.86

AAH5007106

  SDS
alfa aesar™
H5007106
5g Each for $516.82
Description & Specifications

Specifications

Formula Weight 239.51
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Chloropyridine-5-boronic acid pinacol ester
MDL Number MFCD03412793
Synonym 2-Chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
CAS 444120-94-9
Molecular Formula C11H15BClNO2
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.