Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyridine-4-carboxylic acid, 98%

$40.93 - $110.97

Chemical Identifiers

Chemical Name or Material 2-Chloropyridine-4-carboxylic acid
CAS 6313-54-8
Molecular Formula C6H4ClNO2
Formula Weight 157.56
MDL Number MFCD00191402
Beilstein 115861
Synonym 2-Chloroisonicotinic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1944903

 
alfa aesar™
L1944903
1g Each for $40.93

AAL1944906

 
alfa aesar™
L1944906
5g Each for $110.97
Description & Specifications

Specifications

Formula Weight 157.56
Chemical Name or Material 2-Chloropyridine-4-carboxylic acid
Molecular Formula C6H4ClNO2
MDL Number MFCD00191402
CAS 6313-54-8
Assay 98%
Beilstein 115861
Synonym 2-Chloroisonicotinic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 230° to 232°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.