Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyridine-4-carboxamide, 98+%

$531.89 - $531.89

Chemical Identifiers

Chemical Name or Material 2-Chloropyridine-4-carboxamide
CAS 100859-84-5
Molecular Formula C6H5ClN2O
Formula Weight 156.57
MDL Number MFCD00221401
Synonym 2-Chloroisonicotinamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2576906

  SDS
alfa aesar™
H2576906
5g Each for $531.89
Description & Specifications

Specifications

Formula Weight 156.57
Assay ≥98%
MDL Number MFCD00221401
Chemical Name or Material 2-Chloropyridine-4-carboxamide
Molecular Formula C6H5ClN2O
CAS 100859-84-5
Melting Point 201° to 203°C
Synonym 2-Chloroisonicotinamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.