missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyridine-4-boronic acid pinacol ester, 98%

$282.73 - $1,243.11

Chemical Identifiers

CAS 458532-84-8
Molecular Formula C11H15BClNO2
Formula Weight 239.51
MDL Number MFCD04039870
Synonym 2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Chemical Name or Material 2-Chloropyridine-4-boronic acid pinacol ester
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5007003 SDS Alfa Aesar™
H5007003
1g Each for $282.73
AAH5007006 SDS Alfa Aesar™
H5007006
5g Each for $1,243.11
Specifications Description & Specifications

Chemical Identifiers

458532-84-8
239.51
2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
C11H15BClNO2
MFCD04039870
2-Chloropyridine-4-boronic acid pinacol ester

Specifications

1g
C11H15BClNO2
MFCD04039870
2-Chloropyridine-4-boronic acid pinacol ester
458532-84-8
239.51
2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning