Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyridine-4-boronic acid, 95%

$144.69 - $409.45

Chemical Identifiers

Chemical Name or Material 2-Chloropyridine-4-boronic acid
CAS 458532-96-2
Molecular Formula C5H5BClNO2
Formula Weight 157.37
MDL Number MFCD03788416
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2735703

 
alfa aesar™
H2735703
1g Each for $144.69

AAH2735706

 
alfa aesar™
H2735706
5g Each for $409.45
Description & Specifications

Specifications

Formula Weight 157.37
Assay 95%
Recommended Storage 2° to 8°C
Chemical Name or Material 2-Chloropyridine-4-boronic acid
Molecular Formula C5H5BClNO2
MDL Number MFCD03788416
CAS 458532-96-2
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 160° to 164°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C