Please login to your online account to display your discounted pricing

2-chloropyridine-3-sulfonyl chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloropyridine-3-sulfonyl chloride
CAS 6684-06-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% diethylsulfonamide derivative (HPLC)
Assay Percent Range 97%
Molecular Formula C5H3Cl2NO2S
Formula Weight 212.06
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC434310010

 
thermo fisher scientific
AC434310010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC434310050

 
thermo fisher scientific
AC434310050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥96.0% diethylsulfonamide derivative (HPLC)
Formula Weight 212.06
Melting Point 42.0° to 53.0°C
Quantity 1g
Color White to off-white
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Chloropyridine-3-sulfonyl chloride
CAS 6684-06-6
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C5H3Cl2NO2S
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas.
Infrared Spectrum Authentic
Safety and Handling

Danger

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact

GHS P Statement Causes severe skin burns and eye damage. Contact with water liberates toxic gas.