missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloropyridine-3-sulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC434310050

 View more versions of this product

Catalog No. AC434310050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Chloropyridine-3-sulfonyl chloride
Glass bottle
42.0°C to 53.0°C
6684-06-6
100.0
C5H3Cl2NO2S
MFJSIQDQIZEVJX-UHFFFAOYSA-N
2-chloropyridine-3-sulfonyl chloride
10219968
Powder
Authentic
White
5g
96.0
97%
2-chloro-pyridine-3-sulfonyl chloride, 2-chloro-3-pyridinesulfonyl chloride, 2-chloropyridine-3-sulphonyl chloride, 2-chloropyridine-3-sulfonylchloride, 3-pyridinesulfonyl chloride, 2-chloro, 2-chloranylpyridine-3-sulfonyl chloride, 2-chloro-pyridine-3-sulfonyl chlori, pubchem14833, ksc352q1f, 2-chloro-3-chlorosulfonylpyridine
C1=CC(=C(N=C1)Cl)S(=O)(=O)Cl
212.04
212.06
≥96.0% diethylsulfonamide derivative (HPLC)

Chemical Identifiers

6684-06-6
212.04
2-chloro-pyridine-3-sulfonyl chloride, 2-chloro-3-pyridinesulfonyl chloride, 2-chloropyridine-3-sulphonyl chloride, 2-chloropyridine-3-sulfonylchloride, 3-pyridinesulfonyl chloride, 2-chloro, 2-chloranylpyridine-3-sulfonyl chloride, 2-chloro-pyridine-3-sulfonyl chlori, pubchem14833, ksc352q1f, 2-chloro-3-chlorosulfonylpyridine
2-chloropyridine-3-sulfonyl chloride
C5H3Cl2NO2S
MFJSIQDQIZEVJX-UHFFFAOYSA-N
10219968
C1=CC(=C(N=C1)Cl)S(=O)(=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage. Contact with water liberates toxic gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTRE or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

Danger

SDS Documents