missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropyrazine, 98%

Manufacturer:  Alfa Aesar™ A1010806

 View more versions of this product

Catalog No. AAA1010806



Description

Description

Applications
2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.

Solubility
Soluble in Methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

2-Chloropyrazine
153°C to 154°C
14508-49-7
MFCD00006124
107019
GELVZYOEQVJIRR-UHFFFAOYSA-N
2-chloropyrazine
73277
98%
57°C (134°F)
5g
C4H3ClN2
UN1993
chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
C1=CN=C(C=N1)Cl
114.532
114.54

Chemical Identifiers

14508-49-7
114.532
GELVZYOEQVJIRR-UHFFFAOYSA-N
73277
C1=CN=C(C=N1)Cl
C4H3ClN2
MFCD00006124
chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
2-chloropyrazine
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

SDS Documents