missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloropyrazine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 158060500

 View more versions of this product

Catalog No. AC158060500



Description

Description

Specifications

Specifications

2-Chloropyrazine
57°C
Glass bottle
1.28
Colorless to Yellow
14508-49-7
100.0
C4H3ClN2
2-chloro pyrazine, 2-chloro-pyrazine, 2-chloropyrazine, 6-chloropyrazine, chioropyrazine, chloro-pyrazine, chloropyrazin, chloropyrazine, pyrazine, 2-chloro, pyrazine, chloro
GELVZYOEQVJIRR-UHFFFAOYSA-N
2-chloropyrazine
73277
Liquid
1.2800g/mL
Authentic
1.5320 to 1.5390
3% Max. (K.F.)
50g
97.5
97.5% min. (GC)
MFCD00006124
Solubility in water: insoluble in water
ClC1=CN=CC=N1
114.53
114.53
98%

Chemical Identifiers

14508-49-7
114.53
GELVZYOEQVJIRR-UHFFFAOYSA-N
73277
ClC1=CN=CC=N1
C4H3ClN2
MFCD00006124
2-chloro pyrazine, 2-chloro-pyrazine, 2-chloropyrazine, 6-chloropyrazine, chioropyrazine, chloro-pyrazine, chloropyrazin, chloropyrazine, pyrazine, 2-chloro, pyrazine, chloro
2-chloropyrazine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-517-

SDS Documents