Learn More
2-Chloropropionyl chloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 295161000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloropropionyl chloride | |
7623-09-8 | |
100.0 | |
Colorless to Yellow | |
109.0°C to 111.0°C | |
Authentic | |
Glass bottle | |
1.4390 to 1.4410 | |
MFCD00000862 | |
02, 248 | |
2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone | |
JEQDSBVHLKBEIZ-UHFFFAOYNA-N | |
2-chloropropanoyl chloride | |
111019 | |
95% |
95% | |
94.0 | |
-71.0°C | |
1.3000g/mL | |
34°C | |
94% min. (GC) | |
C3H4Cl2O | |
CH3CH(Cl)COCl | |
100 mL | |
1.3 | |
Solubility in water: reacts | |
CC(C(=O)Cl)Cl | |
126.97 | |
126.97 | |
Liquid |
Chemical Identifiers
7623-09-8 | |
126.97 | |
JEQDSBVHLKBEIZ-UHFFFAOYNA-N | |
111019 | |
CC(C(=O)Cl)Cl |
C3H4Cl2O | |
MFCD00000862 | |
2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone | |
2-chloropropanoyl chloride |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or ha
GHS Signal Word: Danger
EINECSNumber : 231-540-7
TSCA : TSCA
RUO – Research Use Only