Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-2-Chloropropionic acid, 94%

Chemical Identifiers

Chemical Name or Material (+/-)-2-Chloropropionic acid
CAS 598-78-7
Molecular Formula C3H5ClO2
Formula Weight 108.53
MDL Number MFCD00004224
UN Number UN2511
Beilstein 1720259
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1286036

  SDS
alfa aesar™
A1286036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.53
Flash Point 101°C (213°F)
RTECS Number UE8575000
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
Quantity 500g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: 2-CHLOROPROPIONIC ACID
Boiling Point 78° to 80°C (11mmHg)
MDL Number MFCD00004224
Health Hazard 1 Danger
Melting Point −13°C
Assay 94%
UN Number UN2511
Beilstein 1720259
TSCA TSCA
Chemical Name or Material (+/-)-2-Chloropropionic acid
CAS 598-78-7
Molecular Formula C3H5ClO2
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.