Please login to your online account to display your discounted pricing

2-chloropropionamide, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC353610500

 View more versions of this product

Catalog No. AC353610500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 107.54
Quantity 50g
Color White to beige
Melting Point 76° to 81°C
Beilstein 02, 249
CAS Max % 100.0
CAS 27816-36-0
Infrared Spectrum Authentic
Chemical Name or Material 2-chloropropionamide
Assay Percent Range 98%
Molecular Formula C3H6ClNO
Linear Formula CH3CH(Cl)CONH
Packaging Glass bottle
MDL Number MFCD00008016
CAS Min % 97.5

Specifications

Chemical Name or Material 2-chloropropionamide
CAS 27816-36-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H6ClNO
Linear Formula CH3CH(Cl)CONH
Formula Weight 107.54
MDL Number MFCD00008016