Please login to your online account to display your discounted pricing

2-chloropropionamide, 98%, ACROS Organics™

$78.09 - $78.09

Chemical Identifiers

Chemical Name or Material 2-chloropropionamide
CAS 27816-36-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H6ClNO
Linear Formula CH3CH(Cl)CONH
Formula Weight 107.54
MDL Number MFCD00008016
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC353610100

  SDS
thermo fisher scientific
353610100
Glass bottle 10g Each for $78.09
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 107.54
Quantity 10g
Color White to beige
Beilstein 02, 249
Melting Point 76° to 81°C
CAS Max % 100.0
Chemical Name or Material 2-chloropropionamide
Packaging Glass bottle
Linear Formula CH3CH(Cl)CONH
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C3H6ClNO
MDL Number MFCD00008016
CAS 27816-36-0
CAS Min % 97.5