Please login to your online account to display your discounted pricing

2-chloropropionamide, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  353610100

 View more versions of this product

Catalog No. AC353610100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 107.54
Physical Form Crystalline Powder
Quantity 10g
Color White to beige
Beilstein 02, 249
Melting Point 76° to 81°C
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula CH3CH(Cl)CONH
Chemical Name or Material 2-chloropropionamide
Assay Percent Range 98%
CAS 27816-36-0
Molecular Formula C3H6ClNO
CAS Min % 97.5
MDL Number MFCD00008016

Specifications

Chemical Name or Material 2-chloropropionamide
CAS 27816-36-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H6ClNO
Linear Formula CH3CH(Cl)CONH
Formula Weight 107.54
MDL Number MFCD00008016