missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropropene, 99%

Manufacturer:  Alfa Aesar™ L1237414

 View more versions of this product

Catalog No. AAL1237414



Description

Description

Applications
2-Chloropropene is used in measurement of photoionization cross sections of allyl and 2-propenyl radicals to form C3H5+ by tunable vacuum ultraviolet synchrotron radiation coupled with photofragment translational spectroscopy.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Store under inert gas. It is sensitive to air. Incompatible with strong oxidizing agents.
Specifications

Specifications

2-Chloropropene
Recognizable
-138°C
557-98-2
MFCD00000859
1361376
PNLQPWWBHXMFCA-UHFFFAOYSA-N
2-chloroprop-1-ene
11203
99%
−34°C (−29°F)
22°C to 23°C
25g
C3H5Cl
UN2456
2-chloropropene, 1-propene, 2-chloro, isopropenyl chloride, 2-chloropropylene, 2-chloro-1-propene, beta-chloropropene, beta-chloropropylene, propene, 2-chloro, unii-50txn55e4a, 2-chloro propylene
CC(=C)Cl
76.523
76.53

Chemical Identifiers

557-98-2
76.523
PNLQPWWBHXMFCA-UHFFFAOYSA-N
11203
CC(=C)Cl
C3H5Cl
MFCD00000859
2-chloropropene, 1-propene, 2-chloro, isopropenyl chloride, 2-chloropropylene, 2-chloro-1-propene, beta-chloropropene, beta-chloropropylene, propene, 2-chloro, unii-50txn55e4a, 2-chloro propylene
2-chloroprop-1-ene
Safety and Handling

Safety and Handling

GHS H Statement
H224-H319-H335
Extremely flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: 2-CHLOROPROPENE

RTECSNumber : UC7200000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents