Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloropropane, 99%

$22.18 - $73.89

Chemical Identifiers

Chemical Name or Material 2-Chloropropane
CAS 75-29-6
Molecular Formula C3H7Cl
Formula Weight 78.54
MDL Number MFCD00000867
UN Number UN2356
Beilstein 1730782
Merck Index 14,5211
Synonym Isopropyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL04535AP

 
alfa aesar™
L04535AP
500mL Each for $73.89

AAL04535AE

 
alfa aesar™
L04535AE
100mL Each for $22.18
Description & Specifications

Specifications

Formula Weight 78.54
Chemical Name or Material 2-Chloropropane
TSCA TSCA
Assay 99%
Molecular Formula C3H7Cl
MDL Number MFCD00000867
CAS 75-29-6
UN Number UN2356
Beilstein 1730782
Merck Index 14,5211
Synonym Isopropyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224-H301-H311-H332
Extremely flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: 2-CHLOROPROPANE
RTECS Number TX4410000
Flash Point −35°C (−31°F)
Odor Slightly Sweetish
Boiling Point 35° to 36°C
Melting Point −117°C
Quantity 500mL

Applications
2-Chloropropane is used as a solvent and an intermediate. It shows anesthetic properties. Further, it is used as an alternative foam-blowing agent. It is used to prepare chlorodiethylisopropylsilane.

Solubility
Miscible with water, ethanol, methanol, benzene and ether.

Notes
Incompatible with strong oxidizing agents, strong bases and metals.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H301-H311-H332
Extremely flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.

2° to 8°C