missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chlorophenylacetylene, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC434750010

 View more versions of this product

Catalog No. AC434750010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-chlorophenylacetylene
Authentic
82°C (16.0 hPa)
1g
97.5
97.5% min. (GC)
MFCD00269951
DGLHLIWXYSGYBI-UHFFFAOYSA-N
1-chloro-2-ethynylbenzene
585995
Liquid
67°C
Glass bottle
Colorless to Yellow
873-31-4
100.0
C8H5Cl
1-chlor-2-ethinylbenzol, 1-chloro-2-ethynyl-benzene, 2'-chlorophenyl acetylene, 2-chlorophenyl acetylene, 2-chlorophenylacetylene, 2;-chlorophenylacetylene, benzene, 1-chloro-2-ethynyl, o-chlorophenylacetylene, o-chlorphenylacetylen, pubchem2550
C#CC1=CC=CC=C1Cl
136.58
136.58
98%

Chemical Identifiers

873-31-4
136.58
DGLHLIWXYSGYBI-UHFFFAOYSA-N
585995
C#CC1=CC=CC=C1Cl
C8H5Cl
MFCD00269951
1-chlor-2-ethinylbenzol, 1-chloro-2-ethynyl-benzene, 2'-chlorophenyl acetylene, 2-chlorophenyl acetylene, 2-chlorophenylacetylene, 2;-chlorophenylacetylene, benzene, 1-chloro-2-ethynyl, o-chlorophenylacetylene, o-chlorphenylacetylen, pubchem2550
1-chloro-2-ethynylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents