missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorophenylacetylene, 98%

Manufacturer:  Alfa Aesar™ H5344106

 View more versions of this product

Catalog No. AAH5344106



Description

Description

Applications
2-Chlorophenylacetylene is used as pharmaceutical intermediate.

Solubility
Soluble in hexane.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications

Specifications

2-Chlorophenylacetylene
82°C (12mmHg)
873-31-4
MFCD00269951
Soluble in hexane.
C#CC1=CC=CC=C1Cl
136.578
136.58
1.125
70°C (158°F)
5g
C8H5Cl
1-chlor-2-ethinylbenzol
DGLHLIWXYSGYBI-UHFFFAOYSA-N
1-chloro-2-ethynylbenzene
585995
98%
1.571

Chemical Identifiers

873-31-4
136.578
DGLHLIWXYSGYBI-UHFFFAOYSA-N
585995
C#CC1=CC=CC=C1Cl
C8H5Cl
MFCD00269951
1-chlor-2-ethinylbenzol
1-chloro-2-ethynylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents