Learn More
2-Chlorophenylacetylene, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5344103
Description
2-Chlorophenylacetylene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chlorophenylacetylene is used as pharmaceutical intermediate.
Solubility
Soluble in hexane.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
2-Chlorophenylacetylene | |
1.125 | |
1 g | |
C8H5Cl | |
2-chlorophenylacetylene, 2'-chlorophenyl acetylene, 1-chloro-2-ethynyl-benzene, benzene, 1-chloro-2-ethynyl, 1-chlor-2-ethinylbenzol, o-chlorphenylacetylen, o-chlorophenylacetylene, pubchem2550, 2-chlorophenyl acetylene, 2;-chlorophenylacetylene | |
DGLHLIWXYSGYBI-UHFFFAOYSA-N | |
1-chloro-2-ethynylbenzene | |
585995 | |
98% |
873-31-4 | |
82°C (12 mmHg) | |
1.571 | |
MFCD00269951 | |
Soluble in hexane. | |
C#CC1=CC=CC=C1Cl | |
136.578 | |
136.58 |
Chemical Identifiers
873-31-4 | |
136.578 | |
DGLHLIWXYSGYBI-UHFFFAOYSA-N | |
585995 | |
C#CC1=CC=CC=C1Cl |
C8H5Cl | |
MFCD00269951 | |
2-chlorophenylacetylene, 2'-chlorophenyl acetylene, 1-chloro-2-ethynyl-benzene, benzene, 1-chloro-2-ethynyl, 1-chlor-2-ethinylbenzol, o-chlorphenylacetylen, o-chlorophenylacetylene, pubchem2550, 2-chlorophenyl acetylene, 2;-chlorophenylacetylene | |
1-chloro-2-ethynylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only