missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorophenyl phosphorodichloridate, 98+%

Manufacturer:  Alfa Aesar™ A1150506

 View more versions of this product

Catalog No. AAA1150506



Specifications

Specifications

2-Chlorophenyl phosphorodichloridate
82°C to 84°C (0.4mmHg)
15074-54-1
MFCD00009704
911009
VLDPXPPHXDGHEW-UHFFFAOYSA-N
1-chloro-2-dichlorophosphoryloxybenzene
84775
≥98%
105°C (221°F)
5g
C6H4Cl3O2P
UN3265
2-chlorophenyl phosphorodichloridate, 2-chlorophenyl dichlorophosphate, o-chlorophenyl dichlorophosphate, phosphorodichloridic acid, 2-chlorophenyl ester, o-chlorophenyl phosphorodichloridate, 2-chlorophenyl phosphoric acid dichloride, 2-chlorophenyl chlorophosphonochloridate, acmc-20anxn, o-chlorophenylphosphonyl dichloride, 2-chlorophenyl dichloridophosphate #
C1=CC=C(C(=C1)OP(=O)(Cl)Cl)Cl
245.42
245.43

Chemical Identifiers

15074-54-1
245.42
VLDPXPPHXDGHEW-UHFFFAOYSA-N
84775
C1=CC=C(C(=C1)OP(=O)(Cl)Cl)Cl
C6H4Cl3O2P
MFCD00009704
2-chlorophenyl phosphorodichloridate, 2-chlorophenyl dichlorophosphate, o-chlorophenyl dichlorophosphate, phosphorodichloridic acid, 2-chlorophenyl ester, o-chlorophenyl phosphorodichloridate, 2-chlorophenyl phosphoric acid dichloride, 2-chlorophenyl chlorophosphonochloridate, acmc-20anxn, o-chlorophenylphosphonyl dichloride, 2-chlorophenyl dichloridophosphate #
1-chloro-2-dichlorophosphoryloxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

SDS Documents