missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chlorophenyl isothiocyanate, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 292240050
Specifications
2-Chlorophenyl isothiocyanate | |
>110°C | |
Glass bottle | |
1.29 | |
Yellow | |
99% | |
98.5 | |
98.5% min. (GC) | |
NCSC6H4Cl | |
12, 601 | |
DASSPOJBUMBXLU-UHFFFAOYSA-N | |
1-chloro-2-isothiocyanatobenzene | |
123171 | |
Liquid |
1.2900g/mL | |
Authentic | |
1.6598 to 1.6618 | |
260°C to 262°C | |
5g | |
2740-81-0 | |
100.0 | |
C7H4ClNS | |
MFCD00004801 | |
1-chloro-2-isothiocyanato-benzene, 2-chlorobenzenisothiocyanate, 2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, acmc-1ccw5, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, isothiocyanic acid 2-chlorophenyl ester, o-chlorophenyl isothiocyanate, o-chlorophenylisothiocyanate | |
C1=CC=C(C(=C1)N=C=S)Cl | |
169.63 | |
169.63 | |
99% |
Chemical Identifiers
2740-81-0 | |
169.63 | |
DASSPOJBUMBXLU-UHFFFAOYSA-N | |
123171 | |
C1=CC=C(C(=C1)N=C=S)Cl |
C7H4ClNS | |
MFCD00004801 | |
1-chloro-2-isothiocyanato-benzene, 2-chlorobenzenisothiocyanate, 2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, acmc-1ccw5, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, isothiocyanic acid 2-chlorophenyl ester, o-chlorophenyl isothiocyanate, o-chlorophenylisothiocyanate | |
1-chloro-2-isothiocyanatobenzene |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash
GHS Signal Word: Danger
TSCA : TSCA