missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorophenoxyacetonitrile, 97%

Manufacturer:  Alfa Aesar™ A1134606

 View more versions of this product

Catalog No. AAA1134606


 

Specifications

Specifications

2-Chlorophenoxyacetonitrile
5g
97%
MFCD00017331
2089865
C1=CC=C(C(=C1)OCC#N)Cl
167.592
167.6
109°C to 110°C (1mmHg)
43111-31-5
C8H6ClNO
UN3276
JWRJGSQLKHGQOT-UHFFFAOYSA-N
2-(2-chlorophenoxy)acetonitrile
521134

Chemical Identifiers

43111-31-5
167.592
JWRJGSQLKHGQOT-UHFFFAOYSA-N
C8H6ClNO
MFCD00017331
521134
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

SDS Documents