Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorophenoxyacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 2-Chlorophenoxyacetonitrile
CAS 43111-31-5
Molecular Formula C8H6ClNO
Formula Weight 167.6
MDL Number MFCD00017331
UN Number UN3276
Beilstein 2089865
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1134606

  SDS
alfa aesar™
A1134606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1134614

  SDS
alfa aesar™
A1134614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1134622

  SDS
alfa aesar™
A1134622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.6
Chemical Name or Material 2-Chlorophenoxyacetonitrile
Assay 97%
Molecular Formula C8H6ClNO
MDL Number MFCD00017331
CAS 43111-31-5
UN Number UN3276
Beilstein 2089865
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Boiling Point 109° to 110°C (1mmHg)
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.